Please login to your online account to display your discounted pricing

Rabbit anti-CD117/c-kit, Clone: R21-V, Novus Biologicals™

$329.44 - $329.44

Specifications

Antigen CD117/c-kit
Clone R21-V
Immunogen Peptide derived from C-terminal sequence of human CD11
Gene Accession No. P10721
Gene ID (Entrez) 3815
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP17901010

 
novus biologicals canada
NBP179010100UL
100μL Each for $329.44
Description & Specifications

Specifications

Antigen CD117/c-kit
Clone R21-V
Immunogen Peptide derived from C-terminal sequence of human CD11
Gene Accession No. P10721
Gene ID (Entrez) 3815
Formulation 20mM Tris-HCl, pH 8. 20mg/mL BSA
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)