Please login to your online account to display your discounted pricing

Rabbit anti-CD117/c-kit, Clone: R21-V, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179010100UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17901010

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Peptide derived from C-terminal sequence of human CD11
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Antigen CD117/c-kit
Format Affinity Purified
Gene Accession No. P10721
Isotype IgG
Purification Method Affinity Purified
Quantity 100μL
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3815
Clone R21-V
Conjugate Unlabeled
Formulation 20mM Tris-HCl, pH 8. 20mg/mL BSA
Monoclonal or Polyclonal Monoclonal