Please login to your online account to display your discounted pricing

Rabbit anti-Calpastatin, Clone: SP82, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen Calpastatin
Clone SP82
Immunogen Synthetic peptide derived from near C-terminus of human calpastatin protein.
Gene ID (Entrez) 831
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21248701

 
novus biologicals canada
NBP21248701ML
0.1mL Each for $309.05

NBP21248705

 
novus biologicals canada
NBP21248705ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen Calpastatin
Clone SP82
Immunogen Synthetic peptide derived from near C-terminus of human calpastatin protein.
Gene ID (Entrez) 831
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)