Please login to your online account to display your discounted pricing

Rabbit anti-Calpastatin, Clone: SP82, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248701

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen Calpastatin
Immunogen Synthetic peptide derived from near C-terminus of human calpastatin protein.
Host Species Rabbit
Cross Reactivity Human
Clone SP82
Reconstitution Protein A purified
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Formulation PBS/1% BSA buffer pH 7.6
Purification Method Purified
Conjugate Unlabeled
Isotype IgG
Quantity 0.1mL
Format Purified
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Bovine, Chicken, Porcine, Rabbit, Rat.
Regulatory Status RUO
Gene ID (Entrez) 831