Please login to your online account to display your discounted pricing

Rabbit anti-Calpastatin, Clone: SP82, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248705

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Antigen Calpastatin
Formulation PBS/1% BSA buffer pH 7.6
Quantity 0.5mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Host Species Rabbit
Isotype IgG
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 831
Test Specificity Predicted to cross react with Bovine, Chicken, Porcine, Rabbit, Rat.
Reconstitution Protein A purified
Clone SP82
Immunogen Synthetic peptide derived from near C-terminus of human calpastatin protein.