Please login to your online account to display your discounted pricing

Rabbit anti-Calpain 1, Clone: SP81, Novus Biologicals™

$309.33 - $551.93

Specifications

Antigen Calpain 1
Clone SP81
Immunogen Synthetic peptide derived from internal region of human calpain protein.
Gene ID (Entrez) 823
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21248605

 
novus biologicals canada
NBP21248605ML
0.5mL Each for $551.93

NBP21248601

 
novus biologicals canada
NBP21248601ML
0.1mL Each for $309.33
Description & Specifications

Specifications

Antigen Calpain 1
Clone SP81
Immunogen Synthetic peptide derived from internal region of human calpain protein.
Gene ID (Entrez) 823
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot