Please login to your online account to display your discounted pricing

Rabbit anti-Calpain 1, Clone: SP81, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248605

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Synthetic peptide derived from internal region of human calpain protein.
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 823
Reconstitution Protein A purified
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Calpain 1
Isotype IgG
Purification Method Purified
Quantity 0.5mL
Monoclonal or Polyclonal Monoclonal
Test Specificity Predicted to cross react with Frog, Mouse, Porcine, Rat.
Clone SP81