Please login to your online account to display your discounted pricing

Rabbit anti-Calpain 1, Clone: SP81, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21248601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21248601

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Rabbit
Isotype IgG
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 823
Monoclonal or Polyclonal Monoclonal
Immunogen Synthetic peptide derived from internal region of human calpain protein.
Quantity 0.1mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Clone SP81
Reconstitution Protein A purified
Purification Method Purified
Antigen Calpain 1
Storage Requirements Store at 4°C. Do not freeze.
Formulation PBS/1% BSA buffer pH 7.6
Test Specificity Predicted to cross react with Frog, Mouse, Porcine, Rat.
Format Purified