Please login to your online account to display your discounted pricing

Rabbit anti-C1qTNF5/CTRP5, Clone: A14-A, Novus Biologicals™

$363.90 - $363.90

Specifications

Antigen C1qTNF5/CTRP5
Clone A14-A
Concentration 2.7mg/mL
Immunogen Peptide close to the amino terminus of human CTRP5
Gene Accession No. Q9BXJ0
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13013105

 
novus biologicals canada
NBP130131005ML
0.05mL Each for $363.90
Description & Specifications

Specifications

Antigen C1qTNF5/CTRP5
Clone A14-A
Concentration 2.7mg/mL
Immunogen Peptide close to the amino terminus of human CTRP5
Gene Accession No. Q9BXJ0
Gene ID (Entrez) 114902
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications ELISA
Applications Western Blotting