Please login to your online account to display your discounted pricing

Rabbit anti-C1qTNF5/CTRP5, Clone: A14-A, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130131005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13013105

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Antigen C1qTNF5/CTRP5
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Gene Accession No. Q9BXJ0
Host Species Rabbit
Applications ELISA
Applications Western Blotting
Immunogen Peptide close to the amino terminus of human CTRP5
Isotype IgG
Purification Method Affinity Purified
Quantity 0.05mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 114902
Test Specificity This antibody is specific for the amino terminus of CTRP5
Clone A14-A
Conjugate Unlabeled
Concentration 2.7mg/mL