Please login to your online account to display your discounted pricing

Rabbit anti-C1qTNF5/CTRP5, Clone: A14-A, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13013101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13013101

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Western Blotting
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 10mg/mL BSA. Immunogen affinity purified.
Host Species Rabbit
Isotype IgG
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody is specific for the amino terminus of CTRP5
Clone A14-A
Cross Reactivity Human
Antigen C1qTNF5/CTRP5
Gene Accession No. Q9BXJ0
Immunogen Peptide close to the amino terminus of human CTRP5
Purification Method Affinity Purified
Regulatory Status RUO
Gene ID (Entrez) 114902
Concentration 2.7mg/mL
Conjugate Unlabeled