missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Rabbit anti-Bovine IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals
Manufacturer: Mp Biomedicals 55274
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Bovine
Specifications
IgG, Whole Molecule | |
Purified | |
IgG | |
Secondary | |
Bovine |
Unconjugated | |
Rabbit | |
5mL | |
Polyclonal |