missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Bovine IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals

$165.66 - $165.66

Specifications

Antigen IgG, Whole Molecule
Form Purified
Host Species Rabbit
Isotype IgG
Primary or Secondary Secondary
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN55267
View Documents Promotion Details
Mp Biomedicals
0855267
2 mL
Each for $165.66
 
Description

Description

Host Species: Rabbit
Isotype: IgG
Species Reactivity: Bovine

Specifications

Specifications

IgG, Whole Molecule
Rabbit
Secondary
Purified
IgG
SDS
Product Certifications