missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Bcl-2, Clone: P21-A, Novus Biologicals™

$307.00 - $307.00

Specifications

Antigen Bcl-2
Clone P21-A
Immunogen peptide derived from the sequence close to N-terminal region of human bcl-2
Gene Accession No. P10415
Gene ID (Entrez) 596
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP17901110 Novus Biologicals Canada
NBP179011100UL
100μL Each for $307.00
Specifications Description & Specifications

Specifications

Bcl-2
peptide derived from the sequence close to N-terminal region of human bcl-2
596
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Immunohistochemistry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin)
P21-A
P10415
20mM Tris-HCl, pH 8.0 Stabilizer: 20mg/mL BSA
Primary
Rabbit
Unlabeled
Affinity Purified