Please login to your online account to display your discounted pricing

Rabbit anti-Bcl-2, Clone: P21-A, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP179011200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP17901120

  • / Each
 

Description & Specifications

Specifications

Antigen Bcl-2
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone P21-A
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 Stabilizer: 20mg/mL BSA
Gene Accession No. P10415
Host Species Rabbit
Immunogen peptide derived from the sequence close to N-terminal region of human bcl-2
Isotype IgG
Purification Method Affinity Purified
Quantity 200μL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 596