Please login to your online account to display your discounted pricing

Rabbit anti-alpha-Methylacyl-CoA Racemase/AMACR, Clone: 13H4, Novus Biologicals™

$142.00 - $417.00

Specifications

Antigen alpha-Methylacyl-CoA Racemase/AMACR
Clone 13H4
Immunogen Full length human recombinant AMACR protein
Gene Accession No. Q9UHK6
Gene ID (Entrez) 23600
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23447101

 
novus biologicals canada
NBP23447101ML
0.1mL Each for $142.00

NBP23447105

 
novus biologicals canada
NBP23447105ML
0.5mL Each for $417.00
Description & Specifications

Specifications

Antigen alpha-Methylacyl-CoA Racemase/AMACR
Clone 13H4
Immunogen Full length human recombinant AMACR protein
Gene Accession No. Q9UHK6
Gene ID (Entrez) 23600
Formulation Culture Medium. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rabbit
Isotype IgG
Conjugate Unlabeled
Format Supernatant
Purification Method Supernatant
Applications Immunohistochemistry (Paraffin)
Applications Western Blot