Please login to your online account to display your discounted pricing

Rabbit anti-alpha-Methylacyl-CoA Racemase/AMACR, Clone: 13H4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23447105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23447105

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Host Species Rabbit
Quantity 0.5mL
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Antigen alpha-Methylacyl-CoA Racemase/AMACR
Gene Accession No. Q9UHK6
Clone 13H4
Gene ID (Entrez) 23600
Immunogen Full length human recombinant AMACR protein
Purification Method Supernatant
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Format Supernatant
Isotype IgG
Formulation Culture Medium. Tissue culture supernatant.