Please login to your online account to display your discounted pricing

Rabbit anti-alpha-Methylacyl-CoA Racemase/AMACR, Clone: 13H4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23447101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23447101

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Antigen alpha-Methylacyl-CoA Racemase/AMACR
Format Supernatant
Formulation Culture Medium. Tissue culture supernatant.
Gene Accession No. Q9UHK6
Host Species Rabbit
Immunogen Full length human recombinant AMACR protein
Isotype IgG
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 23600
Clone 13H4
Conjugate Unlabeled
Purification Method Supernatant
Regulatory Status RUO