Please login to your online account to display your discounted pricing

Rabbit anti-alpha-Fetoprotein/AFP, Clone: SP154, Novus Biologicals™

$309.05 - $551.93

Specifications

Antigen alpha-Fetoprotein/AFP
Clone SP154
Immunogen Purified human Alpha 1 Fetoprotein
Gene ID (Entrez) 174
Formulation PBS/1% BSA buffer pH 7.6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP21251601

 
novus biologicals canada
NBP21251601ML
0.1mL Each for $309.05

NBP21251605

 
novus biologicals canada
NBP21251605ML
0.5mL Each for $551.93
Description & Specifications

Specifications

Antigen alpha-Fetoprotein/AFP
Clone SP154
Immunogen Purified human Alpha 1 Fetoprotein
Gene ID (Entrez) 174
Formulation PBS/1% BSA buffer pH 7.6
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting