Please login to your online account to display your discounted pricing

Rabbit anti-alpha-Fetoprotein/AFP, Clone: SP154, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP21251605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP21251605

  • / Each
 

Description & Specifications

Specifications

Antigen alpha-Fetoprotein/AFP
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone SP154
Conjugate Unlabeled
Format Purified
Formulation PBS/1% BSA buffer pH 7.6
Host Species Rabbit
Immunogen Purified human Alpha 1 Fetoprotein
Isotype IgG
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 174
Test Specificity Predicted to cross react with Bovine, Canine, Porcine.
Reconstitution Protein A purified
Target Species Human