missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rabbit anti-Alkaline Phosphatase/ALPP, Clone: SP15, Novus Biologicals™

$384.00 - $586.39

Specifications

Antigen Alkaline Phosphatase/ALPP
Clone SP15
Immunogen Recombinant protein encoding human placental alkaline phosphatase.
Gene Accession No. P05187
Gene ID (Entrez) 250
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14979501 Novus Biologicals Canada
NBP14979501ML
0.1mL Each for $384.00
NBP14979510 Novus Biologicals Canada
NBP14979510ML
1mL Each for $586.39
Specifications Description & Specifications

Specifications

Alkaline Phosphatase/ALPP
Recombinant protein encoding human placental alkaline phosphatase.
250
Monoclonal
Store at 4°C. Do not freeze.
IgG
Supernatant
Immunohistochemistry,Immunohistochemistry (Paraffin)
SP15
P05187
TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Primary
Rabbit
Unlabeled
Supernatant