Please login to your online account to display your discounted pricing

Rabbit anti-Alkaline Phosphatase/ALPP, Clone: SP15, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979510ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979510

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Recombinant protein encoding human placental alkaline phosphatase.
Isotype IgG
Quantity 1mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 250
Conjugate Unlabeled
Cross Reactivity Human
Antigen Alkaline Phosphatase/ALPP
Format Supernatant
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Gene Accession No. P05187
Purification Method Supernatant
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Clone SP15