Please login to your online account to display your discounted pricing

Rabbit anti-Alkaline Phosphatase/ALPP, Clone: SP15, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14979501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14979501

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Gene Accession No. P05187
Storage Requirements Store at 4°C. Do not freeze.
Format Supernatant
Purification Method Supernatant
Conjugate Unlabeled
Regulatory Status RUO
Primary or Secondary Primary
Isotype IgG
Immunogen Recombinant protein encoding human placental alkaline phosphatase.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SP15
Cross Reactivity Human
Quantity 0.1mL
Formulation TBS/1% BSA buffer pH 7.5. Tissue culture supernatant.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 250
Antigen Alkaline Phosphatase/ALPP