Please login to your online account to display your discounted pricing

Rabbit anti-Alkaline Phosphatase/ALPP, Clone: P16-D, Novus Biologicals™

$354.00 - $719.00

Specifications

Antigen Alkaline Phosphatase/ALPP
Clone P16-D
Immunogen Peptide derived from the middle portion of human PLAP sequence.
Gene Accession No. P05187
Gene ID (Entrez) 250
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13014905

 
novus biologicals canada
NBP13014905ML
0.5mL Each for $440.61

NBP13014910

 
novus biologicals canada
NBP13014910ML
1mL Each for $719.00

NBP22184120

 
novus biologicals canada
NBP221841200UL
200μL Each for $354.00
Description & Specifications

Specifications

Antigen Alkaline Phosphatase/ALPP
Clone P16-D
Immunogen Peptide derived from the middle portion of human PLAP sequence.
Gene Accession No. P05187
Gene ID (Entrez) 250
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rabbit
Cross Reactivity Human
Isotype IgG
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)