Please login to your online account to display your discounted pricing

Rabbit anti-Alkaline Phosphatase/ALPP, Clone: P16-D, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP221841200UL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP22184120

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Conjugate Unlabeled
Cross Reactivity Human
Primary or Secondary Primary
Gene ID (Entrez) 250
Formulation 20mM Tris-HCI, pH 8.0 20mg/mL BSA.
Regulatory Status RUO
Purification Method Affinity Purified
Immunogen Peptide derived from the middle portion of human PLAP sequence.
Format Affinity Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rabbit
Clone P16-D
Quantity 200μL
Gene Accession No. P05187
Antigen Alkaline Phosphatase/ALPP
Monoclonal or Polyclonal Monoclonal