Please login to your online account to display your discounted pricing

Rabbit anti-Alkaline Phosphatase/ALPP, Clone: P16-D, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP13014910ML

 View all options for this product


For Research Use Only

Catalog No. NBP13014910


Description & Specifications


Antigen Alkaline Phosphatase/ALPP
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone P16-D
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 20mM Tris-HCl, pH 8.0 with 20mg/mL BSA.
Gene Accession No. P05187
Host Species Rabbit
Immunogen Peptide derived from the middle portion of human PLAP sequence.
Isotype IgG
Purification Method Affinity Purified
Quantity 1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 250
Test Specificity This antibody is specific to an internal portion of human PLAP sequence.