Thermo Scientific™ RA Lamb Biopsy Inserts
Prevent the loss of small or friable material while allowing interchange of fluids with Thermo Scientific™ RA Lamb Biopsy Inserts.
$303.00 - $303.00
Specifications
Material Stainless Steel
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Color Price Quantity  
Catalog Number Mfr. No. Description Color Price Quantity  
E1087 Thermo Scientific™
E1087
Biopsy insert pad White Case of 100 Each for $303.00
Specifications
Description
Prevent the loss of small or friable material while allowing interchange of fluids with Thermo Scientific™ RA Lamb Biopsy Inserts. These inserts are two-part loose fitting stainless-steel capsules with welded mesh.
  • Mesh fits inside processing cassettes
  • Mesh hole size is 0.5mm
Description & Specifications
Specifications
Stainless Steel