Please login to your online account to display your discounted pricing

Alfa Aesar™ R15K, HIV-1 Inhibitory Peptide

For cellular and molecular biology applications

$68.35 - $87.74

Specifications

Product Type R15K, HIV-1 Inhibitory Peptide
Molecular Weight 1655.97
Storage Requirements -30° to -10°C
Sequence Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66327EXD

 
alfa aesar™
J66327EXD
0.5mg Each for $68.35

AAJ66327MCR

 
alfa aesar™
J66327MCR
1mg Each for $87.74
Description & Specifications

Specifications

Product Type R15K, HIV-1 Inhibitory Peptide
Molecular Weight 1655.97
Storage Requirements -30° to -10°C
Sequence Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys
Format Solid
Regulatory Status RUO
Purification Method HPLC