missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(-)-Tetrahydrofurfurylamine 98.0+%, TCI America™
$96.22 - $924.06
Chemical Identifiers
CAS | 7202-43-9 |
---|---|
Molecular Formula | C5H11NO |
Molecular Weight (g/mol) | 101.149 |
MDL Number | MFCD00192476 |
InChI Key | YNOGYQAEJGADFJ-RXMQYKEDSA-N |
Synonym | r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2-furanmethanamine, tetrahydro-, 2r, 1-2r-tetrahydrofuran-2-yl methanamine, pubchem7001, 2r-tetrahydrofurfurylamine |
PubChem CID | 2734124 |
IUPAC Name | [(2R)-oxolan-2-yl]methanamine |
SMILES | C1CC(OC1)CN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T2551200MG
|
TCI America
T2551200MG |
200 mg |
Each for $96.22
|
|
|||||
T25511G
|
TCI America
T25511G |
1 g |
Each for $287.00
|
|
|||||
T25515G
|
TCI America
T25515G |
5 g |
Each for $924.06
|
|
|||||
Chemical Identifiers
7202-43-9 | |
101.149 | |
YNOGYQAEJGADFJ-RXMQYKEDSA-N | |
2734124 | |
C1CC(OC1)CN |
C5H11NO | |
MFCD00192476 | |
r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2-furanmethanamine, tetrahydro-, 2r, 1-2r-tetrahydrofuran-2-yl methanamine, pubchem7001, 2r-tetrahydrofurfurylamine | |
[(2R)-oxolan-2-yl]methanamine |
Specifications
7202-43-9 | |
60°C | |
Clear Liquid at 20°C | |
MFCD00192476 | |
r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2-furanmethanamine, tetrahydro-, 2r, 1-2r-tetrahydrofuran-2-yl methanamine, pubchem7001, 2r-tetrahydrofurfurylamine | |
C1CC(OC1)CN | |
101.149 | |
101.15 | |
(R)-(-)-Tetrahydrofurfurylamine |
Yellow | |
200 mg | |
C5H11NO | |
2943 | |
YNOGYQAEJGADFJ-RXMQYKEDSA-N | |
[(2R)-oxolan-2-yl]methanamine | |
2734124 | |
≥98.0% (GC,T) |
Safety and Handling
TSCA : No