missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-Tetrahydrofurfurylamine, 98+%, ACROS Organics™

Chemical Identifiers

CAS 7202-43-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00192476
InChI Key YNOGYQAEJGADFJ-RXMQYKEDSA-N
Synonym 1-2r-tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2r, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2r-tetrahydrofurfurylamine, pubchem7001, r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine
PubChem CID 2734124
IUPAC Name [(2R)-oxolan-2-yl]methanamine
SMILES C1CC(OC1)CN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC352630010
SDS View Documents
Thermo Fisher Scientific
AC352630010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

7202-43-9
101.15
YNOGYQAEJGADFJ-RXMQYKEDSA-N
2734124
C1CC(OC1)CN
C5H11NO
MFCD00192476
1-2r-tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2r, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2r-tetrahydrofurfurylamine, pubchem7001, r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine
[(2R)-oxolan-2-yl]methanamine

Specifications

0.9800g/mL
Authentic
1.4540 to 1.456
− 13.50 (20.00°C c−4,MeOH)
151°C to 152°C
1g
98.0
98% min. (GC)
MFCD00192476
Solubility in water: miscible with water
C1CC(OC1)CN
101.15
101.15
98+%
46°C
Glass bottle
0.98
-13.5
Colorless
7202-43-9
100.0
C5H11NO
1-2r-tetrahydrofuran-2-yl methanamine, 2-furanmethanamine, tetrahydro-, 2r, 2r-oxolan-2-ylmethanamine, 2r-tetrahydro-2-furanmethanamine, 2r-tetrahydrofurfurylamine, pubchem7001, r---tetrahydrofurfurylamine, r-tetrahydrofuran-2-yl methanamine, r-tetrahydrofurfurylamine
YNOGYQAEJGADFJ-RXMQYKEDSA-N
[(2R)-oxolan-2-yl]methanamine
2734124
Liquid
(R)-(-)-Tetrahydrofurfurylamine, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Warning

SDS