Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-Tetrahydrofurfurylamine, 98+%

Chemical Identifiers

Chemical Name or Material (R)-(-)-Tetrahydrofurfurylamine
CAS 7202-43-9
Molecular Formula C5H11NO
Formula Weight 101.15
MDL Number MFCD00192476
UN Number UN2943
Beilstein 4652873
Synonym (R)-(-)-2-(Aminomethyl)tetrahydrofuran
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1441003

  SDS
alfa aesar™
L1441003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL14410MD

  SDS
alfa aesar™
L14410MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 101.15
Chemical Name or Material (R)-(-)-Tetrahydrofurfurylamine
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 7202-43-9
MDL Number MFCD00192476
Assay ≥98%
UN Number UN2943
Quantity 1g
Molecular Formula C5H11NO
Health Hazard 1 Danger
Synonym (R)-(-)-2-(Aminomethyl)tetrahydrofuran
Optical Rotation −12° (c=2 in Chloroform)
Flash Point 45°C (113°F)
Boiling Point 152° to 153°C
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Beilstein 4652873
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TETRAHYDROFURFURYLAMINE

Applications
(R)-(-)-Tetrahydrofurfurylamine is used in the synthesis of novel topoisomerase I targeting anti-cancer agents with mild to potent cytotoxic activity. Also used in the discovery and synthesis of orally bioavailable stearoyl-CoA desaturase 1 inhibitors.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.