missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-Tetrahydrofurfuryl alcohol, 98+%

$94.53 - $255.99

Chemical Identifiers

CAS 22415-59-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD03093085
InChI Key BSYVTEYKTMYBMK-RXMQYKEDSA-N
Synonym r-tetrahydrofurfuryl alcohol, r-tetrahydrofuran-2-yl methanol, 2-furanmethanol, tetrahydro-, 2r, 2r-oxolan-2-yl methanol, r---tetrahydrofurfuryl alcohol, 2r-oxolan-2-ylmethanol, r---tetrahydrofurfurylalcohol, 2r-tetrahydrofuran-2-yl methanol, 2r-2-oxolanyl methanol, r-tetrahydrofuran-2-methanol
PubChem CID 6931745
IUPAC Name [(2R)-oxolan-2-yl]methanol
SMILES C1CC(OC1)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL19044MD View Documents Alfa Aesar™
L19044MD
250mg Each for $94.53
 
AAL1904403 View Documents Alfa Aesar™
L1904403
1g Each for $255.99
 
Specifications

Chemical Identifiers

22415-59-4
102.133
BSYVTEYKTMYBMK-RXMQYKEDSA-N
6931745
C1CC(OC1)CO
C5H10O2
MFCD03093085
r-tetrahydrofurfuryl alcohol, r-tetrahydrofuran-2-yl methanol, 2-furanmethanol, tetrahydro-, 2r, 2r-oxolan-2-yl methanol, r---tetrahydrofurfuryl alcohol, 2r-oxolan-2-ylmethanol, r---tetrahydrofurfurylalcohol, 2r-tetrahydrofuran-2-yl methanol, 2r-2-oxolanyl methanol, r-tetrahydrofuran-2-methanol
[(2R)-oxolan-2-yl]methanol

Specifications

75°C (167°F)
173°C to 175°C
22415-59-4
MFCD03093085
r-tetrahydrofurfuryl alcohol, r-tetrahydrofuran-2-yl methanol, 2-furanmethanol, tetrahydro-, 2r, 2r-oxolan-2-yl methanol, r---tetrahydrofurfuryl alcohol, 2r-oxolan-2-ylmethanol, r---tetrahydrofurfurylalcohol, 2r-tetrahydrofuran-2-yl methanol, 2r-2-oxolanyl methanol, r-tetrahydrofuran-2-methanol
C1CC(OC1)CO
102.133
102.13
(R)-(-)-Tetrahydrofurfuryl alcohol
−2.3° (Neat)
250mg
C5H10O2
79846
BSYVTEYKTMYBMK-RXMQYKEDSA-N
[(2R)-oxolan-2-yl]methanol
6931745
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H319-H227
Causes serious eye irritation.
Combustible liquid.

GHS P Statement
P210-P280-P305+P351+P338-P337+P313-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning