Please login to your online account to display your discounted pricing

(R)-(+)-Tetrahydro-2-furoic acid, 99% (98% ee), ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-(+)-Tetrahydro-2-furoic acid
Name Note 99% (98% e.e.)
CAS 87392-05-0
Assay ≥98.5%
Molecular Formula C5H8O3
Formula Weight 116.12
MDL Number MFCD00211271
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369500010

  SDS
thermo fisher scientific
AC369500010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 116.12
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.2100g/mL
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00211271
Name Note 99% (98% e.e.)
Molecular Formula C5H8O3
Chemical Name or Material (R)-(+)-Tetrahydro-2-furoic acid
CAS 87392-05-0
Health Hazard 1 GHS Signal Word: Danger
Specific Rotation +18° to +20° (20°C, 589nm) (neat)
Packaging Glass bottle
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.