Learn More
(R)-Styrene oxide, 95%, 95% ee, Thermo Scientific Chemicals
$91.67 - $265.88
Chemical Identifiers
CAS | 20780-53-4 |
---|---|
Molecular Formula | C8H8O |
Molecular Weight (g/mol) | 120.15 |
MDL Number | MFCD00066210 |
InChI Key | AWMVMTVKBNGEAK-QMMMGPOBSA-N |
Synonym | r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r |
PubChem CID | 114705 |
ChEBI | CHEBI:45389 |
IUPAC Name | (2R)-2-phenyloxirane |
SMILES | C1O[C@@H]1C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC240550010
|
Thermo Scientific Chemicals
240550010 |
1 g | Glass bottle |
Each for $91.67
|
|
||||
AC240550050
|
Thermo Scientific Chemicals
240550050 |
5 g | Glass bottle |
Each for $265.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20780-53-4 | |
120.15 | |
AWMVMTVKBNGEAK-QMMMGPOBSA-N | |
114705 | |
(2R)-2-phenyloxirane |
C8H8O | |
MFCD00066210 | |
r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r | |
CHEBI:45389 | |
C1O[C@@H]1C1=CC=CC=C1 |
Specifications
20780-53-4 | |
100.0 | |
Yellow | |
192.0°C to 195.0°C | |
Authentic | |
Glass bottle | |
1.5320 to 1.5360 | |
1 g | |
−21° (23°C c=5,chloroform) | |
r-styrene oxide, r-phenyloxirane, r-+-styrene oxide, r-2-phenyloxirane, 2r-2-phenyloxirane, r-epoxyethyl benzene, oxirane, phenyl-, 2r, benzene, epoxyethyl-, r | |
AWMVMTVKBNGEAK-QMMMGPOBSA-N | |
(2R)-2-phenyloxirane | |
114705 | |
120.15 | |
Liquid |
95 | |
-37.0°C | |
1.0530g/mL | |
79°C | |
94% min. (GC) | |
C8H8O | |
MFCD00066210 | |
1.053 | |
− 21.00 | |
Solubility in water: practically insoluble. Other solubilities: soluble in hexane,petroleum ether,acetone,,alcohol,and benzene | |
C1O[C@@H]1C1=CC=CC=C1 | |
120.15 | |
CHEBI:45389 | |
95%,95% ee | |
(R)-Styrene oxide |
Safety and Handling
GHS H Statement
May cause cancer.
May cause genetic defects.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Danger
RUO – Research Use Only