Please login to your online account to display your discounted pricing

(R)-(+)-sec-Phenethylealcohol, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-(+)-sec-Phenethyl alcohol
CAS 1517-69-7
CAS Min % 98.5
CAS Max % 100.0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula C6H5CH(CH3)OH
Formula Weight 122.17g/mol
MDL Number MFCD00064263
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC217780050

  SDS
thermo fisher scientific
217780050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Specific Rotation +41° to +44° (20°C, 589nm) (neat)
Refractive Index 1.5260 to 1.5280 (20°C, 589nm)
Density 1.0100g/mL
Specific Rotation Condition + 42.50 (20.00°C neat)
Color Clear colorless
Quantity 5g
Chemical Name or Material (R)-(+)-sec-Phenethyl alcohol
CAS 1517-69-7
CAS Min % 98.5
CAS Max % 100.0
Physical Form Clear colorless liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C8H10O
Linear Formula C6H5CH(CH3)OH
Formula Weight 122.17g/mol
MDL Number MFCD00064263
Beilstein 06, IV, 3029
Synonym (R)-(+)-α-Methylbenzyl alcohol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Chiral derivatization reagent

Safety and Handling

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.