Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-S-Methyl-S-phenylsulfoximine, 97%

$117.14 - $352.50

Chemical Identifiers

Chemical Name or Material (R)-(-)-S-Methyl-S-phenylsulfoximine
CAS 60933-65-5
Molecular Formula C7H9NOS
Formula Weight 155.22
MDL Number MFCD00151460
Beilstein 5245635
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0856203

 
alfa aesar™
L0856203
1g Each for $352.50

AAL08562MD

 
alfa aesar™
L08562MD
250mg Each for $117.14
Description & Specifications

Specifications

Formula Weight 155.22
Chemical Name or Material (R)-(-)-S-Methyl-S-phenylsulfoximine
Assay 97%
Molecular Formula C7H9NOS
MDL Number MFCD00151460
CAS 60933-65-5
Beilstein 5245635
Melting Point 29° to 31°C
Quantity 1g
Optical Rotation −36° (c=1 in Acetone)

Applications
(R)-(-)-S-Methyl-S-phenylsulfoximine is used as chiral ligands.

Notes
Hygroscopic. Store away from water and oxidizing agents. Protect from humidity and water. Incompatible with strong oxidizing agents.