Please login to your online account to display your discounted pricing

Alfa Aesar™ [(R,R)-TsDPEN-Ru(mesitylene)Cl]

Chemical Identifiers

Chemical Name or Material [(R,R)-TsDPEN-Ru(mesitylene)Cl]
CAS 174813-82-2
Formula Weight 622.18
MDL Number MFCD03791031
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3799303

  SDS
alfa aesar™
H3799303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material [(R,R)-TsDPEN-Ru(mesitylene)Cl]
CAS 174813-82-2
Physical Form Powder
Formula Weight 622.18
MDL Number MFCD03791031
Color Brown
Quantity 1g