Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II)

Chemical Identifiers

Chemical Name or Material (R,R)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II)
CAS 192139-92-7
Molecular Formula C31H35ClN2O2RuS
Formula Weight 636.21
MDL Number MFCD09953466
Synonym [(R,R)-TsDPEN Ru(p-cymene)Cl]; (R,R)-Tosyl-DPEN-(p-cymene)ruthenium chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25943MD

 
alfa aesar™
H25943MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (R,R)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II)
CAS 192139-92-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C31H35ClN2O2RuS
Formula Weight 636.21
MDL Number MFCD09953466
Synonym [(R,R)-TsDPEN Ru(p-cymene)Cl]; (R,R)-Tosyl-DPEN-(p-cymene)ruthenium chloride
Quantity 250mg