Learn More
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 295810250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride | |
97.5 | |
96.0 % min. | |
97% | |
C36H52ClMnN2O2 | |
25g | |
1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. | |
CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] | |
635.21 | |
635.21 |
138124-32-0 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD02101664 | |
15, 5300 | |
YRVXCOIDMFNGIJ-SEILFYAJSA-M | |
2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride | |
131675872 |
Chemical Identifiers
138124-32-0 | |
635.21 | |
YRVXCOIDMFNGIJ-SEILFYAJSA-M | |
131675872 | |
CC(C)(C)C1=CC(=CNC2CCCCC2NC=C3C=C(C=C(C3=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C.[Cl-].[Mn] |
C36H52ClMnN2O2 | |
MFCD02101664 | |
1r,2r---1,2-cyclohexanediamino-n,n'-bis 3,5-di-t-butylsalicylidene manganese iii chloride r,r-jacobsen cat. | |
2,4-ditert-butyl-6-[[[(1R,2R)-2-[(3,5-ditert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methylamino]cyclohexyl]amino]methylidene]cyclohexa-2,4-dien-1-one;manganese;chloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO â Research Use Only