Please login to your online account to display your discounted pricing

Alfa Aesar™ [(R,R)-MsDpen-Ru(p-cymene)Cl]

$311.11 - $383.66

Chemical Identifiers

Chemical Name or Material [(R,R)-MsDpen-Ru(p-cymene)Cl]
CAS 300664-92-0
Molecular Formula C25H31ClN2O2RuS
Formula Weight 560.12g/mol
MDL Number MFCD12546180
Synonym (R,R)-Mesyl-DPEN-(p-cymene)ruthenium chloride; (R,R)-N-Methylsulfonyl-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3001603

 
alfa aesar™
H30016-03
1g Each for $383.66

AAH30016MD

 
alfa aesar™
H30016-MD
250mg Each for $311.11
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material [(R,R)-MsDpen-Ru(p-cymene)Cl]
CAS 300664-92-0
Molecular Formula C25H31ClN2O2RuS
Formula Weight 560.12g/mol
MDL Number MFCD12546180
Synonym (R,R)-Mesyl-DPEN-(p-cymene)ruthenium chloride; (R,R)-N-Methylsulfonyl-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.