missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R,R)-DIPAMP, 90%, Thermo Scientific Chemicals
$145.79 - $761.93
Chemical Identifiers
CAS | 55739-58-7 |
---|---|
Molecular Formula | C28H28O2P2 |
Molecular Weight (g/mol) | 458.48 |
MDL Number | MFCD05863546 |
InChI Key | QKZWXPLBVCKXNQ-UHFFFAOYNA-N |
Synonym | r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane |
PubChem CID | 10884975 |
SMILES | COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH28374MC
|
Thermo Scientific Chemicals
H28374MC |
100 mg |
Each for $145.79
|
|
|||||
AAH28374MD
|
Thermo Scientific Chemicals
H28374MD |
250 mg |
Each for $271.10
|
|
|||||
AAH2837403
|
Thermo Scientific Chemicals
H2837403 |
1 g |
Each for $761.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
55739-58-7 | |
458.48 | |
QKZWXPLBVCKXNQ-UHFFFAOYNA-N | |
10884975 |
C28H28O2P2 | |
MFCD05863546 | |
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane | |
COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1 |
Specifications
55739-58-7 | |
C28H28O2P2 | |
100 mg | |
QKZWXPLBVCKXNQ-UHFFFAOYNA-N | |
COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1 | |
458.48 | |
458.47 | |
(R,R)-DIPAMP |
101°C to 105°C | |
MFCD05863546 | |
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane | |
−83° (c=1 in Chloroform) | |
(R)-(2-methoxyphenyl)-[2-[(2-methoxyphenyl)-phenylphosphanyl]ethyl]-phenylphosphane | |
10884975 | |
97% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only