missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R,R)-DIPAMP, 90%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H28374MD

 View more versions of this product

Catalog No. AAH28374MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R,R)-DIPAMP
101°C to 105°C
MFCD05863546
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
−83° (c=1 in Chloroform)
(2-methoxyphenyl)({2-[(2-methoxyphenyl)(phenyl)phosphanyl]ethyl})phenylphosphane
10884975
97%
55739-58-7
C28H28O2P2
250 mg
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1
458.48
458.47

Chemical Identifiers

55739-58-7
458.48
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
10884975
C28H28O2P2
MFCD05863546
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only