missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine, 97+%

$160.38 - $575.22

Chemical Identifiers

CAS 799297-44-2
Molecular Formula C21H22NP
Molecular Weight (g/mol) 319.388
MDL Number MFCD17018768
InChI Key JWZAIGGNEGTDMG-UTKZUKDTSA-N
Synonym 1r,2r-1-diphenylphosphino-1-phenylpropan-2-amine, 1r,2r-2-amino-1-phenylpropyl diphenylphosphane, 1r,2r-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1r,2r-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1r,2r-1-diphenylphosphino-1-phenyl-2-propanamine, r,r---2-amino-1-phenylpropyldiphenylphosphine
PubChem CID 11982345
IUPAC Name (1R,2R)-1-diphenylphosphanyl-1-phenylpropan-2-amine
SMILES CC(C(C1=CC=CC=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60639MD View Documents Alfa Aesar™
H60639MD
250mg Each for $160.38
 
AAH6063903 View Documents Alfa Aesar™
H6063903
1g Each for $575.22
 
Specifications

Chemical Identifiers

799297-44-2
319.388
JWZAIGGNEGTDMG-UTKZUKDTSA-N
11982345
CC(C(C1=CC=CC=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)N
C21H22NP
MFCD17018768
1r,2r-1-diphenylphosphino-1-phenylpropan-2-amine, 1r,2r-2-amino-1-phenylpropyl diphenylphosphane, 1r,2r-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1r,2r-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1r,2r-1-diphenylphosphino-1-phenyl-2-propanamine, r,r---2-amino-1-phenylpropyldiphenylphosphine
(1R,2R)-1-diphenylphosphanyl-1-phenylpropan-2-amine

Specifications

−140° (c=0.5 in Chloroform)
250mg
C21H22NP
UN3259
JWZAIGGNEGTDMG-UTKZUKDTSA-N
(1R,2R)-1-diphenylphosphanyl-1-phenylpropan-2-amine
11982345
≥97%
105°C to 109°C
799297-44-2
MFCD17018768
1r,2r-1-diphenylphosphino-1-phenylpropan-2-amine, 1r,2r-2-amino-1-phenylpropyl diphenylphosphane, 1r,2r-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1r,2r-1-diphenylphosphanyl-1-phenylpropan-2-amine, 1r,2r-1-diphenylphosphino-1-phenyl-2-propanamine, r,r---2-amino-1-phenylpropyldiphenylphosphine
CC(C(C1=CC=CC=C1)P(C2=CC=CC=C2)C3=CC=CC=C3)N
319.388
319.38
(R,R)-(-)-2-Amino-1-phenylpropyldiphenylphosphine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.