Please login to your online account to display your discounted pricing

(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine, 96%, ACROS Organics™


Quantity 100mg
Packaging Glass Ampule with Screw-cap

Manufacturer: thermo fisher scientific  AC293451000

 View more versions of this product

Catalog No. AC293451000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 159.23
Quantity 100mg
Color Clear colorless to light yellow
Packaging Glass Ampule with Screw-cap
TSCA TSCA
Specific Rotation Condition − 8.00 (20.00°C c=3,EtOH)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Specific Rotation -7° to -9° (20°C, 589nm) (c=3, ethanol)
Infrared Spectrum Authentic
Name Note 96%
Molecular Formula C8H17NO2
CAS 90290-05-4
Chemical Name or Material (R, R)-(-)-2, 5-Bis(methoxymethyl)pyrrolidine
Refractive Index 1.4470 to 1.4500 (20°C, 589nm)

Specifications

Chemical Name or Material (R, R)-(-)-2, 5-Bis(methoxymethyl)pyrrolidine
Name Note 96%
CAS 90290-05-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H17NO2
Formula Weight 159.23