Please login to your online account to display your discounted pricing

(R,R)-(-)-2,5-Bis(methoxymethyl)pyrrolidine, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R, R)-(-)-2, 5-Bis(methoxymethyl)pyrrolidine
Name Note 96%
CAS 90290-05-4
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C8H17NO2
Formula Weight 159.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC293451000

  SDS
thermo fisher scientific
AC293451000
Glass Ampule with Screw-cap 100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 159.23
Quantity 100mg
Color Clear colorless to light yellow
Packaging Glass Ampule with Screw-cap
TSCA TSCA
Specific Rotation Condition − 8.00 (20.00°C c=3,EtOH)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Specific Rotation -7° to -9° (20°C, 589nm) (c=3, ethanol)
Infrared Spectrum Authentic
Name Note 96%
Molecular Formula C8H17NO2
CAS 90290-05-4
Chemical Name or Material (R, R)-(-)-2, 5-Bis(methoxymethyl)pyrrolidine
Refractive Index 1.4470 to 1.4500 (20°C, 589nm)