Please login to your online account to display your discounted pricing

(R,R)-(+)-1,2-Diphenyl-1,2-ethanediol, ≥98%, ACROS Organics™

$65.24 - $65.24

Chemical Identifiers

Chemical Name or Material (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol
CAS 52340-78-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C14H14O2
Linear Formula C6H5CH(OH)CH(OH)C6H5
Formula Weight 214.26
MDL Number MFCD00064254
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC279525000

  SDS
thermo fisher scientific
279525000
Glass bottle 500mg Each for $65.24
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Crystalline Powder or Crystals
Formula Weight 214.26
Quantity 500mg
Color White to beige or light brown
Merck Index 14, 4777
Beilstein 06, 970
TSCA TSCA
Melting Point 146° to 150°C
Specific Rotation Condition +93° (20°C c=2.5,EtOH)
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Specific Rotation +92° to +94° (20°C, 589nm) (c=2.5, EtOH)
Packaging Glass bottle
CAS 52340-78-0
Chemical Name or Material (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol
Linear Formula C6H5CH(OH)CH(OH)C6H5
MDL Number MFCD00064254
Assay Percent Range 98+%
Molecular Formula C14H14O2