missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R,R)-(+)-1,2-Diphenyl-1,2-ethanediol, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 279525000
Specifications
(R,R)-(+)-1,2-Diphenyl-1,2-ethanediol | |
Glass bottle | |
+ 93.00 | |
146.0°C to 150.0°C | |
52340-78-0 | |
100.0 | |
C14H14O2 | |
MFCD00064254 | |
14, 4777 | |
IHPDTPWNFBQHEB-ZIAGYGMSSA-N | |
(1R,2R)-1,2-diphenylethane-1,2-diol | |
853019 | |
214.26 | |
98+% |
Authentic | |
+93° (20°C c=2.5,EtOH) | |
Beige to White or Brown | |
500mg | |
98.0 | |
98% min. (GC) | |
C6H5CH(OH)CH(OH)C6H5 | |
06, 970 | |
+-1r,2r-1,2-diphenylethane-1,2-diol, +-dihydrobenzoin, +-hydrobenzoin, 1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel, 1r,2r-1,2-diphenylethane-1,2-diol, isohydrobenzoin, r,r-+-hydrobenzoin, r,r-hydrobenzoin, unii-wv0i33nond, wv0i33nond | |
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O | |
214.26 | |
CHEBI:50014 | |
Crystalline Powder or Crystals |
Chemical Identifiers
52340-78-0 | |
214.26 | |
IHPDTPWNFBQHEB-ZIAGYGMSSA-N | |
853019 | |
(1R,2R)-1,2-diphenylethane-1,2-diol |
C14H14O2 | |
MFCD00064254 | |
+-1r,2r-1,2-diphenylethane-1,2-diol, +-dihydrobenzoin, +-hydrobenzoin, 1,2-ethanediol, 1,2-diphenyl-, 1r,2r-rel, 1r,2r-1,2-diphenylethane-1,2-diol, isohydrobenzoin, r,r-+-hydrobenzoin, r,r-hydrobenzoin, unii-wv0i33nond, wv0i33nond | |
CHEBI:50014 | |
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O |
Safety and Handling
TSCA : TSCA