Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Di(4-pyridyl)-1,2-ethanediamine tetrahydrochloride, 95%, ee 99%

Chemical Identifiers

Chemical Name or Material (R,R)-1,2-Di(4-pyridyl)-1,2-ethanediamine tetrahydrochloride
Molecular Formula C12H14N4·4HCl
Formula Weight 360.11
MDL Number MFCD09265313
Synonym (R,R)-1,2-Bis(4-pyridyl)-1,2-ethanediamine tetrahydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH25967MD

 
alfa aesar™
H25967MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material (R,R)-1,2-Di(4-pyridyl)-1,2-ethanediamine tetrahydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Assay 95%
Assay Percent Notes ee 99%
Molecular Formula C12H14N4·4HCl
Formula Weight 360.11
MDL Number MFCD09265313
Synonym (R,R)-1,2-Bis(4-pyridyl)-1,2-ethanediamine tetrahydrochloride
Quantity 250mg