Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Di(2-pyridyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$826.59 - $826.59

Chemical Identifiers

Chemical Name or Material (R,R)-1,2-Di(2-pyridyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C12H14N4·2HCl
Formula Weight 287.2
MDL Number MFCD09265311
Synonym (R,R)-1,2-Bis(2-pyridyl)-1,2-ethanediamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2597403

  SDS
alfa aesar™
H2597403
1g Each for $826.59
Description & Specifications

Specifications

Formula Weight 287.2
Chemical Name or Material (R,R)-1,2-Di(2-pyridyl)-1,2-ethanediamine dihydrochloride
Assay 95%
Assay Percent Notes ee 99%
Molecular Formula C12H14N4·2HCl
MDL Number MFCD09265311
Synonym (R,R)-1,2-Bis(2-pyridyl)-1,2-ethanediamine dihydrochloride
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.