Please login to your online account to display your discounted pricing

Alfa Aesar™ (R,R)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

$248.62 - $248.62

Chemical Identifiers

Chemical Name or Material (R,R)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride
Molecular Formula C22H20N2·2HCl
Formula Weight 385.35
MDL Number MFCD09265310
Synonym (R,R)-1,2-Bis(1-naphthyl)-1,2-ethanediamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2598903

  SDS
alfa aesar™
H2598903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH25989MD

  SDS
alfa aesar™
H25989MD
250mg Each for $248.62
Description & Specifications

Specifications

Formula Weight 385.35
Synonym (R,R)-1,2-Bis(1-naphthyl)-1,2-ethanediamine dihydrochloride
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Molecular Formula C22H20N2·2HCl
Assay Percent Notes ee 99%
Assay 95%
MDL Number MFCD09265310
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material (R,R)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.